Gene Transcripts

Gene Symbol
SPICE1
Entrez Gene ID
152185
Gene Name
spindle and centriole associated protein 1
Gene Aliases
CCDC52, SPICE
Chromosome Location
Chr. 3: 113442718 - 113515187 on Build GRCh38
UniGene ID
Hs.477144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144718.3 NP_653319.1 16 2635
XM_011512461.2 XP_011510763.1 16 2392
XM_017005775.1 XP_016861264.1 16 2392
XM_017005776.1 XP_016861265.1 16 2529
XM_017005777.1 XP_016861266.1 16 2452
XM_017005778.1 XP_016861267.1 16 2636
GenBank mRNA AK129575.1 - 6 1164
AL832148.1 - 16 2604
AL832327.2 - 16 4300
AY099107.1 - 16 2555
BC019232.1 - 2 131
BC036951.1 - 16 2576 5500345
BC038595.1 - 4 758

View Full Product Details