Gene Transcripts

Gene Symbol
KIAA1549L
Entrez Gene ID
25758
Gene Name
KIAA1549 like
Gene Aliases
C11orf41, C11orf69, G2
Chromosome Location
Chr. 11: 33376569 - 33674102 on Build GRCh38
UniGene ID
Hs.502266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012194.2 NP_036326.2 1 1269
XM_005252847.3 XP_005252904.2 3 1345
XM_005252848.3 XP_005252905.1 1 1269
XM_011519969.2 XP_011518271.1 1 1269
XM_017017484.1 XP_016872973.1 1 1269
XM_017017485.1 XP_016872974.1 1 1269
GenBank mRNA AK124395.1 - 1 1269
U10991.1 - 1 673

View Full Product Details