Gene Transcripts

Gene Symbol
C22orf24
Entrez Gene ID
25775
Gene Name
chromosome 22 open reading frame 24
Gene Aliases
HSN44A4A
Chromosome Location
Chr. 22: 31933521 - 31945362 on Build GRCh38
UniGene ID
Hs.627602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302819.1 NP_001289748.1 2 1442
NM_001302820.1 NP_001289749.1 2 1369
NM_015372.2 NP_056187.1 3 1233
NR_126507.1 3 1160
GenBank mRNA AL050256.1 - 3 1221

View Full Product Details