Gene Transcripts
- Gene Symbol
- TEX40
- Entrez Gene ID
- 25858
- Gene Name
- testis expressed 40
- Gene Aliases
- C11orf20
- Chromosome Location
- Chr. 11: 64300391 - 64304767 on Build GRCh38
- UniGene ID
- Hs.97174
- Species
- Homo sapiens
- Gene Symbol
- KCNK4-TEX40
- Entrez Gene ID
- 106780802
- Gene Name
- KCNK4-TEX40 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 11: 64291722 - 64304769 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_133662.1 |
|
11 |
2596 |
|
View Full Product Details