Gene Transcripts

Gene Symbol
TMEM74
Entrez Gene ID
157753
Gene Name
transmembrane protein 74
Gene Aliases
NET36
Chromosome Location
Chr. 8: 108606850 - 108787615 on Build GRCh38
UniGene ID
Hs.99439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153015.2 NP_694560.1 2 961
GenBank mRNA AK055230.1 - 2 887
BC030710.2 - 2 961 4819093

View Full Product Details