Gene Transcripts

Gene Symbol
FAM47A
Entrez Gene ID
158724
Gene Name
family with sequence similarity 47 member A
Gene Aliases
Chromosome Location
Chr. X: 34129752 - 34132330 on Build GRCh38
UniGene ID
Hs.143268
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203408.3 NP_981953.2 1 2106
GenBank mRNA AK292354.1 - 2 2051
BC026171.2 - 2 2051 4827969

View Full Product Details