Gene Transcripts

Gene Symbol
RHOXF1
Entrez Gene ID
158800
Gene Name
Rhox homeobox family member 1
Gene Aliases
OTEX, PEPP1
Chromosome Location
Chr. X: 120109049 - 120117651 on Build GRCh38
UniGene ID
Hs.644617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139282.2 NP_644811.1 3 561
XM_011531281.2 XP_011529583.1 4 612
GenBank mRNA AY099086.1 - 3 561
BC069324.1 - 3 536 7262157
BC069529.1 - 3 538 7262169
BC103857.1 - 3 520 40003132
BC103858.1 - 3 520 40003136

View Full Product Details