Gene Transcripts

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
Chromosome Location
Chr. 12: 88142296 - 88199887 on Build GRCh38
UniGene ID
Hs.331268
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181783.3 NP_861448.2 14 2417
XM_011537980.2 XP_011536282.1 13 2192
XM_011537981.2 XP_011536283.1 7 1081
GenBank mRNA AJ697717.1 - 14 2287
AK074973.1 - 13 2044
AK290436.1 - 14 2417
AK302755.1 - 14 2417
BC117175.1 - 13 2238 40125726
BC117177.1 - 13 2239 40125728
BX538169.1 - 14 2390
CR749244.1 - 14 2369
CR749309.1 - 14 2404
CR933632.1 - 14 2368
CR936640.1 - 4 745

View Full Product Details