Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256404.1 | NP_001243333.1 | 16 | 2870 | |
NM_198459.3 | NP_940861.3 | 13 | 2330 | ||
GenBank mRNA | BX647950.1 | - | 13 | 2331 | |
BX649075.1 | - | 13 | 1456 | ||
CR749576.1 | - | 16 | 2794 |