Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289000.1 | NP_001275929.1 | 4 | 437 | |
NM_001289001.1 | NP_001275930.1 | 4 | 447 | ||
NM_153609.3 | NP_705837.1 | 4 | 516 | ||
XM_011529989.2 | XP_011528291.1 | 4 | 575 | ||
GenBank mRNA | AJ319876.1 | - | 3 | 379 | |
AY055384.1 | - | 4 | 516 | ||
AY358398.1 | - | 4 | 447 | ||
BC039082.1 | - | 4 | 519 | 5164881 | |
CR456446.1 | - | 4 | 437 |