Gene Transcripts
- Gene Symbol
- GRAMD2
- Entrez Gene ID
- 196996
- Gene Name
- GRAM domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 15: 72159806 - 72197814 on Build GRCh38
- UniGene ID
- Hs.596332
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012642.2 |
NP_001012660.1
|
3, 4 |
216 |
|
XM_011521327.2 |
XP_011519629.1
|
3, 4 |
325 |
|
XM_011521328.2 |
XP_011519630.1
|
3, 4 |
676 |
|
XM_011521329.2 |
XP_011519631.1
|
3, 4 |
193 |
|
XM_011521330.2 |
XP_011519632.1
|
3, 4 |
277 |
|
XM_011521331.2 |
XP_011519633.1
|
3, 4 |
216 |
|
XM_011521332.2 |
XP_011519634.1
|
4, 5 |
400 |
|
XM_011521333.2 |
XP_011519635.1
|
4, 5 |
417 |
|
XM_017021997.1 |
XP_016877486.1
|
3, 4 |
360 |
|
XM_017021998.1 |
XP_016877487.1
|
3, 4 |
584 |
|
GenBank mRNA |
AK095072.1 |
- |
3, 4 |
206 |
|
BC038451.1 |
- |
3, 4 |
190 |
|
DA854662.1 |
- |
3, 4 |
216 |
|
View Full Product Details