Gene Transcripts
- Gene Symbol
- NLRC3
- Entrez Gene ID
- 197358
- Gene Name
- NLR family CARD domain containing 3
- Gene Aliases
- CLR16.2, NOD3
- Chromosome Location
- Chr. 16: 3538272 - 3578373 on Build GRCh38
- UniGene ID
- Hs.592091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178844.3 |
NP_849172.2
|
19, 20 |
3531 |
|
NR_075083.2 |
|
20, 21 |
3728 |
|
XM_017023027.1 |
XP_016878516.1
|
19, 20 |
3561 |
|
XM_017023028.1 |
XP_016878517.1
|
20, 21 |
3735 |
|
XM_017023029.1 |
XP_016878518.1
|
21, 22 |
3816 |
|
XM_017023030.1 |
XP_016878519.1
|
21, 22 |
3877 |
|
XM_017023031.1 |
XP_016878520.1
|
19, 20 |
3622 |
|
XM_017023032.1 |
XP_016878521.1
|
21, 22 |
4578 |
|
XM_017023033.1 |
XP_016878522.1
|
21, 22 |
3755 |
|
XM_017023034.1 |
XP_016878523.1
|
20, 21 |
3674 |
|
XM_017023035.1 |
XP_016878524.1
|
20, 21 |
3703 |
|
XM_017023036.1 |
XP_016878525.1
|
19, 20 |
4262 |
|
XM_017023037.1 |
XP_016878526.1
|
20, 21 |
3456 |
|
XM_017023038.1 |
XP_016878527.1
|
20, 21 |
3429 |
|
GenBank mRNA |
AK074109.1 |
- |
15, 16 |
1409 |
|
AK090431.1 |
- |
20, 21 |
3726 |
|
AY601811.1 |
- |
19, 20 |
3518 |
|
FJ889357.1 |
- |
16, 17 |
3107 |
|
View Full Product Details