Gene Transcripts

Gene Symbol
EPHA1
Entrez Gene ID
2041
Gene Name
EPH receptor A1
Gene Aliases
EPH, EPHT, EPHT1
Chromosome Location
Chr. 7: 143390813 - 143408892 on Build GRCh38
UniGene ID
Hs.89839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005232.4 NP_005223.4 8 1570
XM_006715880.3 XP_006715943.1 7 1198
GenBank mRNA AK290351.1 - 8 1534
AK313263.1 - 8 1543
BC130291.1 - 8 1554 40146322
BC144156.1 - 7 1706
CD653381.1 - 3 459
M18391.1 - 8 1576
Z27409.1 - 4 628

View Full Product Details