Gene Transcripts

Gene Symbol
RAB12
Entrez Gene ID
201475
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 18: 8609428 - 8639383 on Build GRCh38
UniGene ID
Hs.270074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025300.2 NP_001020471.2 3, 4 711
XM_006722300.3 XP_006722363.1 3, 4 1024
GenBank mRNA AK127824.1 - 2, 3 326

View Full Product Details