Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
Chromosome Location
Chr. 9: 114603897 - 114646423 on Build GRCh38
UniGene ID
Hs.522357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153045.3 NP_694590.2 4 500
XM_006716997.1 XP_006717060.1 4 503
XM_011518350.1 XP_011516652.1 5 671
XM_011518351.2 XP_011516653.1 5 799
XM_011518352.1 XP_011516654.1 5 668
XM_011518353.1 XP_011516655.1 5 671
XM_011518354.1 XP_011516656.1 4 706
XM_011518355.1 XP_011516657.1 4 703
XM_017014429.1 XP_016869918.1 5 827
XM_017014430.1 XP_016869919.1 4 703
GenBank mRNA AK289383.1 - 4 469
AL557166.3 - 4 604
AL834412.1 - 2 112
BC107133.1 - 4 438 40025594
BC107134.1 - 4 651 40025600
BI553818.1 - 4 509
BQ049157.1 - 4 569
BX649023.1 - 4 386

View Full Product Details