Gene Transcripts

Gene Symbol
NAIF1
Entrez Gene ID
203245
Gene Name
nuclear apoptosis inducing factor 1
Gene Aliases
C9orf90, bA379C10.2
Chromosome Location
Chr. 9: 128061233 - 128067321 on Build GRCh38
UniGene ID
Hs.373606
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197956.3 NP_931045.1 2 1519
GenBank mRNA AK122729.1 - 2 1519
AL833392.1 - 2 1311

View Full Product Details