Gene Transcripts

Gene Symbol
ZCCHC5
Entrez Gene ID
203430
Gene Name
zinc finger CCHC-type containing 5
Gene Aliases
Mar3, Mart3, ZHC5
Chromosome Location
Chr. X: 78656069 - 78659328 on Build GRCh38
UniGene ID
Hs.134873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152694.2 NP_689907.1 2 993
GenBank mRNA AK096184.1 - 2 993
BC136548.1 - 1 901 9020538
BC136549.1 - 1 901 9020539

View Full Product Details