Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166261.1 | NP_001159733.1 | 3 | 454 | |
NM_001166262.1 | NP_001159734.1 | 3 | 524 | ||
NM_001166263.1 | NP_001159735.1 | 4 | 553 | ||
NM_152912.4 | NP_690876.3 | 3 | 492 | ||
XM_006719771.3 | XP_006719834.1 | 4 | 540 | ||
XM_006719772.3 | XP_006719835.1 | 3 | 771 | ||
XM_011534957.2 | XP_011533259.1 | 4 | 1014 | ||
XM_011534958.2 | XP_011533260.1 | 4 | 798 | ||
XM_011534959.2 | XP_011533261.1 | 4 | 963 | ||
XM_011534960.2 | XP_011533262.1 | 4 | 628 | ||
XM_011534961.2 | XP_011533263.1 | 4 | 1216 | ||
XM_011534962.2 | XP_011533264.1 | 4 | 793 | ||
XM_011534963.2 | XP_011533265.1 | 4 | 1381 | ||
XM_017020417.1 | XP_016875906.1 | 3 | 398 | ||
GenBank mRNA | AF410851.1 | - | 2 | 359 | |
AK094420.1 | - | 1 | 1553 | ||
BC039599.1 | - | 3 | 445 | 3855096 | |
BC046166.1 | - | 3 | 464 | 3460180 | |
BG617536.1 | - | 3 | 493 | ||
BI460170.1 | - | 4 | 549 | ||
BM548061.1 | - | 2 | 417 | ||
CN272769.1 | - | 4 | 477 |