Gene Transcripts

Gene Symbol
EPS8
Entrez Gene ID
2059
Gene Name
epidermal growth factor receptor pathway substrate 8
Gene Aliases
DFNB102
Chromosome Location
Chr. 12: 15620141 - 15789576 on Build GRCh38
UniGene ID
Hs.591160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004447.5 NP_004438.3 17 2240
XM_011520605.2 XP_011518907.1 18 2300
GenBank mRNA AK291777.1 - 17 2045
AK292931.1 - 17 1897
AK301834.1 - 10 1230
AK304860.1 - 17 2240
AK316134.1 - 10 1149
AK316239.1 - 10 1422
BC030010.2 - 17 2052 4820933
U12535.1 - 17 2012

View Full Product Details