Gene Transcripts

Gene Symbol
SLC16A9
Entrez Gene ID
220963
Gene Name
solute carrier family 16 member 9
Gene Aliases
C10orf36, MCT9
Chromosome Location
Chr. 10: 59650764 - 59710079 on Build GRCh38
UniGene ID
Hs.499709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323977.1 NP_001310906.1 4 328
NM_001323978.1 NP_001310907.1 5 756
NM_001323979.1 NP_001310908.1 4 443
NM_001323980.1 NP_001310909.1 4 673
NM_001323981.1 NP_001310910.1 5 886
NM_194298.2 NP_919274.1 4 803
XM_017015883.1 XP_016871372.1 4 803
XM_017015884.1 XP_016871373.1 5 1136
GenBank mRNA AK125791.1 - 4 463
BC126238.1 - 3 392 8991954
BC130567.1 - 3 392 40146598
DA759951.1 - 4 328

View Full Product Details