Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 3301448 - 4269000 on Build GRCh38
UniGene ID
Hs.653013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079653.1 NP_001073121.1 7 1200
NM_152744.3 NP_689957.3 33 5055
XM_011515188.1 XP_011513490.1 27 3893
XM_011515189.1 XP_011513491.1 27 3893
XM_011515190.1 XP_011513492.1 27 3893
XM_017011837.1 XP_016867326.1 27 3893
XM_017011838.1 XP_016867327.1 27 3893
XM_017011839.1 XP_016867328.1 27 3893
GenBank mRNA AL833354.1 - 7 1197

View Full Product Details