Gene Transcripts

Gene Symbol
RSBN1L
Entrez Gene ID
222194
Gene Name
round spermatid basic protein 1 like
Gene Aliases
Chromosome Location
Chr. 7: 77696426 - 77779803 on Build GRCh38
UniGene ID
Hs.592289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198467.2 NP_940869.2 7 1913
GenBank mRNA AK124517.1 - 3 280
AK127713.1 - 5 1108
BC059402.1 - 7 1884 30347338

View Full Product Details