Gene Transcripts

Gene Symbol
SPESP1
Entrez Gene ID
246777
Gene Name
sperm equatorial segment protein 1
Gene Aliases
ESP, SP-ESP
Chromosome Location
Chr. 15: 68930500 - 68946811 on Build GRCh38
UniGene ID
Hs.657932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145658.3 NP_663633.1 2 1141
GenBank mRNA AF275321.1 - 2 1045
AY358983.1 - 2 1124
BC017998.1 - 2 1115 4252566

View Full Product Details