Gene Transcripts

Gene Symbol
EPHX4
Entrez Gene ID
253152
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7, EH4, EPHXRP
Chromosome Location
Chr. 1: 92029976 - 92063536 on Build GRCh38
UniGene ID
Hs.201555
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173567.4 NP_775838.3 3 483
GenBank mRNA AK074822.1 - 3 443
BC041475.1 - 3 458 5244314

View Full Product Details