Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322278.1 | NP_001309207.1 | 4 | 563 | |
NM_001322279.1 | NP_001309208.1 | 4 | 527 | ||
NM_199227.2 | NP_954697.1 | 4 | 477 | ||
XM_017003750.1 | XP_016859239.1 | 4 | 476 | ||
GenBank mRNA | AY374142.1 | - | 4 | 477 | |
BC113644.1 | - | 4 | 422 | 8322696 | |
BC143913.1 | - | 4 | 508 | ||
DQ005576.1 | - | 4 | 791 |