Gene Transcripts

Gene Symbol
CXorf58
Entrez Gene ID
254158
Gene Name
chromosome X open reading frame 58
Gene Aliases
Chromosome Location
Chr. X: 23907932 - 23939520 on Build GRCh38
UniGene ID
Hs.351265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK058173.1 - 2 423
BC117344.1 - 2 335 40125895
BC117346.1 - 2 335 40125897
BC144007.1 - 2 335 9052526

View Full Product Details