Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207340.1 | NP_997223.1 | 3 | 1115 | |
GenBank mRNA | AK301787.1 | - | 3 | 1149 | |
AY358448.1 | - | 3 | 1115 | ||
BC005015.1 | - | 2 | 529 | ||
BC057833.1 | - | 3 | 944 | 30343331 |