Gene Transcripts

Gene Symbol
MORN5
Entrez Gene ID
254956
Gene Name
MORN repeat containing 5
Gene Aliases
C9orf113, C9orf18
Chromosome Location
Chr. 9: 122159661 - 122200088 on Build GRCh38
UniGene ID
Hs.71428
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286828.1 NP_001273757.1 3 315
NM_198469.3 NP_940871.2 4 427
XM_005251877.4 XP_005251934.1 4 423
XM_005251878.3 XP_005251935.1 4 398
XM_006717034.3 XP_006717097.1 3 363
XM_011518457.2 XP_011516759.1 4 423
XM_011518458.1 XP_011516760.1 5 506
XM_017014578.1 XP_016870067.1 2 414
GenBank mRNA AK128877.1 - 4 424
AK302081.1 - 4 401
BC133690.1 - 4 418 40119769
BC143440.1 - 3 275
HY011325.1 - 4 428

View Full Product Details