Gene Transcripts

Gene Symbol
RASGEF1C
Entrez Gene ID
255426
Gene Name
RasGEF domain family member 1C
Gene Aliases
Chromosome Location
Chr. 5: 180100795 - 180209130 on Build GRCh38
UniGene ID
Hs.190559
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175062.3 NP_778232.2 4 502
GenBank mRNA AF449764.1 - 3 690
AK093160.1 - 1 354
BC036802.1 - 5 520
BC057759.1 - 1 156 5268021
BF967301.1 - 5 525

View Full Product Details