Gene Transcripts
- Gene Symbol
- WDR72
- Entrez Gene ID
- 256764
- Gene Name
- WD repeat domain 72
- Gene Aliases
- AI2A3
- Chromosome Location
- Chr. 15: 53513741 - 53762878 on Build GRCh38
- UniGene ID
- Hs.122125
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_182758.3 |
NP_877435.3
|
16, 17 |
2912 |
|
NR_102334.1 |
|
16, 17 |
3110 |
|
XM_011521433.2 |
XP_011519735.1
|
17, 18 |
3109 |
|
XM_011521435.2 |
XP_011519737.1
|
16, 17 |
2963 |
|
XM_011521436.2 |
XP_011519738.1
|
16, 17 |
2938 |
|
XM_011521437.2 |
XP_011519739.1
|
15, 16 |
2836 |
|
XM_017022061.1 |
XP_016877550.1
|
16, 17 |
2907 |
|
GenBank mRNA |
AK125130.1 |
- |
2, 3 |
523 |
|
BC101614.1 |
- |
15, 16 |
2891 |
8069120 |
BC101616.1 |
- |
15, 16 |
2891 |
8069122 |
BX537884.1 |
- |
16, 17 |
2912 |
|
BX648571.1 |
- |
16, 17 |
3112 |
|
BX648751.1 |
- |
10, 11 |
2293 |
|
View Full Product Details