Gene Transcripts

Gene Symbol
TAS2R40
Entrez Gene ID
259286
Gene Name
taste 2 receptor member 40
Gene Aliases
GPR60, T2R40, T2R58
Chromosome Location
Chr. 7: 143222079 - 143223050 on Build GRCh38
UniGene ID
Hs.352241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176882.1 NP_795363.1 1 260
GenBank mRNA BC101795.1 - 1 302
BC101797.1 - 1 302 8069303

View Full Product Details