Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320289.1 | NP_001307218.1 | 9 | 1308 | |
NM_001320290.1 | NP_001307219.1 | 9 | 1045 | ||
NM_178033.1 | NP_828847.1 | 9 | 1342 | ||
XM_017000973.1 | XP_016856462.1 | 9 | 1373 | ||
GenBank mRNA | AK091806.1 | - | 9 | 1046 | |
AK098065.1 | - | 9 | 1335 | ||
AK131355.1 | - | 9 | 1308 | ||
AM040940.1 | - | 9 | 1179 | ||
AY358537.1 | - | 9 | 1325 | ||
BC028102.1 | - | 9 | 1342 | 5243005 |