Gene Transcripts

Gene Symbol
BLOC1S2
Entrez Gene ID
282991
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
BLOS2, BORCS2, CEAP, CEAP11
Chromosome Location
Chr. 10: 100273278 - 100286712 on Build GRCh38
UniGene ID
Hs.34906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001342.2 NP_001001342.1 2 323
NM_001282436.1 NP_001269365.1 4 388
NM_001282437.1 NP_001269366.1 3 399
NM_001282438.1 NP_001269367.1 3 250
NM_001282439.1 NP_001269368.1 3 280
NM_173809.4 NP_776170.2 3 280
XM_017016102.1 XP_016871591.1 3 379
GenBank mRNA AB593084.1 - 3 242
AK054697.1 - 2 316
AK298969.1 - 3 248
AY278457.1 - 2 278
AY531265.1 - 3 227
BC020494.1 - 2 151 3900723
BF666010.1 - 4 355
BG545838.1 - 2 318
BG679165.1 - 3 252
BI753469.1 - 3 240
CN340962.1 - 3 367
DB456773.1 - 3 248
HY107885.1 - 2 325
HY165508.1 - 3 251

View Full Product Details