Gene Transcripts

Gene Symbol
SLC39A5
Entrez Gene ID
283375
Gene Name
solute carrier family 39 member 5
Gene Aliases
LZT-Hs7, MYP24, ZIP5
Chromosome Location
Chr. 12: 56230036 - 56237846 on Build GRCh38
UniGene ID
Hs.591018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135195.1 NP_001128667.1 11 1938
NM_173596.2 NP_775867.2 13 1967
XM_005268803.1 XP_005268860.1 11 1855
XM_011538198.1 XP_011536500.1 11 1920
XM_011538199.1 XP_011536501.1 13 1974
XM_011538200.1 XP_011536502.1 12 1924
XM_011538201.1 XP_011536503.1 12 1898
XM_017019185.1 XP_016874674.1 10 1707
XM_017019186.1 XP_016874675.1 13 2107
XM_017019187.1 XP_016874676.1 11 1856
GenBank mRNA AK172768.1 - 11 1901
BC027884.1 - 11 1812 5221329

View Full Product Details