Gene Transcripts

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
Chromosome Location
Chr. 12: 42081845 - 42144871 on Build GRCh38
UniGene ID
Hs.259347
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099650.1 NP_001093120.1 6 1243
NM_173601.1 NP_775872.1 7 1336
XM_017019211.1 XP_016874700.1 7 1075
GenBank mRNA AK125141.1 - 6 1096
BC015597.2 - 3 398
BC030023.1 - 7 1344 5266607
BC039145.1 - 6 1083 4508300

View Full Product Details