Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
Chromosome Location
Chr. 1: 169511954 - 169586632 on Build GRCh38
UniGene ID
Hs.30054
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000130.4 NP_000121.2 13 3271
XM_017000660.1 XP_016856149.1 12 3280
GenBank mRNA AJ297255.1 - 1 1151
AK226079.1 - 13 3189
M14335.1 - 13 3202
M16967.1 - 13 3216

View Full Product Details