Gene Transcripts

Gene Symbol
LOC102724813
Entrez Gene ID
102724813
Gene Name
protein FRG1B
Gene Aliases
Chromosome Location
Chr. 9|NT_187375: 3219 - 27031 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011546190.2 XP_011544492.1 5 527
XM_017030110.1 XP_016885599.1 4 601

View Full Product Details