Gene Transcripts

Gene Symbol
ZADH2
Entrez Gene ID
284273
Gene Name
zinc binding alcohol dehydrogenase domain containing 2
Gene Aliases
Chromosome Location
Chr. 18: 75195108 - 75209348 on Build GRCh38
UniGene ID
Hs.465433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306093.1 NP_001293022.1 2 556
NM_175907.5 NP_787103.1 2 734
GenBank mRNA AK292880.1 - 2 489
AK302801.1 - 2 286
BC018081.1 - 2 712
BC033780.1 - 2 620 4510603
BC078661.1 - 2 547 5575156
BX361234.2 - 2 736
BX647730.1 - 2 557

View Full Product Details