Gene Transcripts

Gene Symbol
NWD1
Entrez Gene ID
284434
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
Chromosome Location
Chr. 19: 16719526 - 16817963 on Build GRCh38
UniGene ID
Hs.406014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290355.1 NP_001277284.1 20 5441
XM_011527929.1 XP_011526231.1 20 4973
XM_011527930.2 XP_011526232.1 20 5459
XM_011527931.2 XP_011526233.1 19 5239
XM_011527934.1 XP_011526236.1 15 3178
XM_017026669.1 XP_016882158.1 21 5526
GenBank mRNA AK090685.1 - 2 407
BC117698.1 - 18 4437 40073616
BX648940.1 - 21 5441

View Full Product Details