Gene Transcripts
- Gene Symbol
- C3orf33
- Entrez Gene ID
- 285315
- Gene Name
- chromosome 3 open reading frame 33
- Gene Aliases
- AC3-33
- Chromosome Location
- Chr. 3: 155762612 - 155806287 on Build GRCh38
- UniGene ID
- Hs.350846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308229.1 |
NP_001295158.1
|
2, 3 |
207 |
|
NM_173657.2 |
NP_775928.1
|
3, 4 |
334 |
|
XM_005247379.4 |
XP_005247436.1
|
2, 3 |
1179 |
|
XM_011512711.2 |
XP_011511013.1
|
2, 3 |
311 |
|
GenBank mRNA |
AF115515.1 |
- |
2, 3 |
170 |
|
AK055701.1 |
- |
3, 4 |
313 |
|
AK289890.1 |
- |
2, 3 |
207 |
|
AL832165.1 |
- |
2, 3 |
192 |
|
BC042198.1 |
- |
2, 3 |
169 |
|
BC064898.1 |
- |
2, 3 |
179 |
6008321 |
DA275323.1 |
- |
2, 3 |
207 |
|
View Full Product Details