Gene Transcripts
- Gene Symbol
- SUMF1
- Entrez Gene ID
- 285362
- Gene Name
- sulfatase modifying factor 1
- Gene Aliases
- AAPA3037, FGE, UNQ3037
- Chromosome Location
- Chr. 3: 4034714 - 4467282 on Build GRCh38
- UniGene ID
- Hs.350475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164674.1 |
NP_001158146.1
|
7, 8 |
984 |
|
NM_182760.3 |
NP_877437.2
|
8, 9 |
1059 |
|
GenBank mRNA |
AB448737.1 |
- |
8, 9 |
1030 |
|
AK057983.1 |
- |
9, 10 |
958 |
|
AK075459.1 |
- |
8, 9 |
1041 |
|
AK295240.1 |
- |
4, 5 |
650 |
|
AK302018.1 |
- |
7, 8 |
952 |
|
AY208752.1 |
- |
8, 9 |
1022 |
|
AY323910.1 |
- |
8, 9 |
1022 |
|
BC017005.2 |
- |
3, 4 |
298 |
|
BC110862.1 |
- |
8, 9 |
1030 |
5760490 |
BC121122.2 |
- |
8, 9 |
1051 |
40120670 |
View Full Product Details