Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207352.3 | NP_997235.3 | 7 | 1244 | |
XM_005262935.3 | XP_005262992.1 | 7 | 1029 | ||
XM_017008037.1 | XP_016863526.1 | 6 | 1002 | ||
GenBank mRNA | AK122600.1 | - | 7 | 979 | |
AK126473.1 | - | 8 | 913 | ||
AY422002.1 | - | 7 | 1244 | ||
BC060857.1 | - | 7 | 1110 | 30333559 | |
BX648730.1 | - | 3 | 1150 |