Gene Transcripts

Gene Symbol
GPRIN3
Entrez Gene ID
285513
Gene Name
GPRIN family member 3
Gene Aliases
GRIN3
Chromosome Location
Chr. 4: 89237666 - 89308010 on Build GRCh38
UniGene ID
Hs.100912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198281.2 NP_938022.2 2 2294
XM_005262936.3 XP_005262993.1 2 2534
XM_005262937.4 XP_005262994.1 2 2588
XM_017008043.1 XP_016863532.1 2 4371
XM_017008044.1 XP_016863533.1 2 2092
GenBank mRNA AB095947.2 - 1 2859
AK124616.1 - 2 2077
BC126371.1 - 1 1908 8992087

View Full Product Details