Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173829.3 | NP_776190.1 | 3 | 258 | |
XM_011543344.2 | XP_011541646.1 | 2 | 390 | ||
GenBank mRNA | AK094073.1 | - | 258 | ||
AL832348.1 | - | 3 | 215 | ||
BC047006.1 | - | 3 | 187 | ||
BC108716.1 | - | 3 | 188 | 6184559 | |
BC127085.1 | - | 3 | 196 | 40122236 | |
BC127086.1 | - | 3 | 196 | 40122239 |