Gene Transcripts

Gene Symbol
KRT73
Entrez Gene ID
319101
Gene Name
keratin 73
Gene Aliases
IRT6IRS3, K6IRS3, KRT6IRS3
Chromosome Location
Chr. 12: 52601813 - 52620133 on Build GRCh38
UniGene ID
Hs.55410
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175068.2 NP_778238.1 4 819
GenBank mRNA AJ508776.1 - 4 819
BC109212.2 - 4 809 40007200

View Full Product Details