Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204883.1 | NP_001191812.1 | 4 | 1151 | |
NM_001204884.1 | NP_001191813.1 | 4 | 1139 | ||
NM_001204885.1 | NP_001191814.1 | 4 | 1129 | ||
NM_001204886.1 | NP_001191815.1 | 4 | 1166 | ||
NM_001204887.1 | NP_001191816.1 | 4 | 1511 | ||
NM_001204888.1 | NP_001191817.1 | 3 | 1403 | ||
NM_198490.2 | NP_940892.1 | 3 | 1417 | ||
GenBank mRNA | AK128345.1 | - | 3 | 1213 | |
AY166852.2 | - | 3 | 1395 | ||
BC062319.1 | - | 3 | 1253 | 4873224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204890.1 | NP_001191819.1 | 13 | 2004 | |
GenBank mRNA | AB032986.1 | - | 13 | 1810 |