Gene Transcripts

Gene Symbol
FAM92B
Entrez Gene ID
339145
Gene Name
family with sequence similarity 92 member B
Gene Aliases
Chromosome Location
Chr. 16: 85098352 - 85112508 on Build GRCh38
UniGene ID
Hs.125713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198491.2 NP_940893.1 8 836
XM_005255931.2 XP_005255988.1 8 836
XM_011523063.1 XP_011521365.1 8 836
XM_017023198.1 XP_016878687.1 8 836
GenBank mRNA AK126284.1 - 8 836
BC093665.1 - 8 718 7939510
BC111944.1 - 8 718 8327412

View Full Product Details