Gene Transcripts
- Gene Symbol
- MROH2A
- Entrez Gene ID
- 339766
- Gene Name
- maestro heat like repeat family member 2A
- Gene Aliases
- HEATR7B1
- Chromosome Location
- Chr. 2: 233774022 - 233833423 on Build GRCh38
- UniGene ID
- Hs.361778
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287395.1 |
NP_001274324.1
|
37, 38 |
4573 |
|
XM_011511075.2 |
XP_011509377.1
|
37, 38 |
4780 |
|
XM_011511076.2 |
XP_011509378.1
|
37, 38 |
4768 |
|
XM_011511077.2 |
XP_011509379.1
|
37, 38 |
4750 |
|
XM_011511078.2 |
XP_011509380.1
|
37, 38 |
4741 |
|
XM_011511079.2 |
XP_011509381.1
|
37, 38 |
4735 |
|
XM_011511080.2 |
XP_011509382.1
|
37, 38 |
6251 |
|
XM_011511081.2 |
XP_011509383.1
|
36, 37 |
4675 |
|
XM_011511082.2 |
XP_011509384.1
|
36, 37 |
4642 |
|
XM_011511083.2 |
XP_011509385.1
|
37, 38 |
4780 |
|
XM_011511084.2 |
XP_011509386.1
|
36, 37 |
4582 |
|
XM_011511085.2 |
XP_011509387.1
|
31, 32 |
3850 |
|
XM_011511086.2 |
XP_011509388.1
|
28, 29 |
3505 |
|
GenBank mRNA |
BC042133.1 |
- |
2, 3 |
400 |
|
View Full Product Details