Gene Transcripts

Gene Symbol
TMPRSS11A
Entrez Gene ID
339967
Gene Name
transmembrane protease, serine 11A
Gene Aliases
ECRG1
Chromosome Location
Chr. 4: 67910301 - 67963514 on Build GRCh38
UniGene ID
Hs.450110
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114387.1 NP_001107859.1 9 1178
NM_182606.3 NP_872412.3 9 1187
GenBank mRNA AF071882.1 - 9 1138
AK131518.1 - 9 1187
AK292950.1 - 9 1116
AY498712.1 - 9 1090
BC111796.1 - 9 1104 40026103

View Full Product Details