Gene Transcripts

Gene Symbol
AKR1C8P
Entrez Gene ID
340811
Gene Name
aldo-keto reductase family 1 member C8, pseudogene
Gene Aliases
AKR1CL1
Chromosome Location
Chr. 10: 5154460 - 5185187 on Build GRCh38
UniGene ID
Hs.282513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027916.2 2 244
GenBank mRNA AK131335.1 - 2 246
BC101204.1 - 2 238 40021857
BC101205.1 - 2 238 40021860

View Full Product Details